Assay Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034858.3 | NP_001030030.2 | 13 - 14 | 2102 | 51 | |
| GenBank mRNA | AK131686.1 | - | 9 - 10 | 1299 | 51 | |
| AK141039.1 | - | 7 - 8 | 2434 | 51 | ||
| BC172168.1 | - | 8 - 9 | 1844 | 51 | ||
More Information
Set Membership: |
|

Armc2
Genomic Map