Assay Details
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011317.4 | NP_035447.3 | 2 - 3 | 651 | 100 | |
NR_045036.1 | 2 - 3 | 651 | 100 | |||
GenBank mRNA | AK044927.1 | - | 2 - 3 | 647 | 100 | |
AK050520.1 | - | 2 - 3 | 646 | 100 | ||
AK150687.1 | - | 2 - 3 | 630 | 100 | ||
AK152084.1 | - | 2 - 3 | 630 | 100 | ||
BC002051.1 | - | 2 - 3 | 607 | 3490667 | 100 | |
U17046.1 | - | 2 - 3 | 608 | 100 | ||
U17961.1 | - | 2 - 3 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
