Assay Details
| Gene Symbol: Il23r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144548.1 | NP_653131.2 | 1 - 2 | 85 | 92 | |
| GenBank mRNA | AF461423.1 | - | 1 - 2 | 85 | 92 | |
| BC112425.1 | - | 1 - 2 | 68 | 40062206 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF461423.1, BC112425.1, NM_144548.1 transcripts.
Set Membership: |
|

Il23r
Genomic Map