Assay Details
Gene Symbol: Fancd2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027633.3 | NP_081909.2 | 3 - 4 | 766 | 121 | |
GenBank mRNA | AK016461.1 | - | 3 - 4 | 707 | 121 | |
BC042706.1 | - | 1 - 2 | 422 | 121 | ||
BC049660.1 | - | 3 - 4 | 766 | 6742399 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760447 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760445 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760446 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | NM_027633.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | AK016461.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | BC042706.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | BC049660.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | NM_027633.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | AK016461.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | BC042706.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P410228 | BC049660.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46459 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718251 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13981 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34663 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106689 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | NM_027633.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | AK016461.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | BC042706.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | BC049660.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | NM_027633.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | AK016461.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | BC042706.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P410228 | BC049660.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760442 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760422 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760445 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760444 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760434 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06031494 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106689 | - | - |
Whole Mouse Genome G4122A | A_51_P410228 | NM_027633.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410228 | AK016461.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410228 | BC042706.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410228 | BC049660.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410228 | NM_027633.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410228 | AK016461.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410228 | BC042706.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410228 | BC049660.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135269 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106689 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05151700 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05190820 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05151704 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05620341 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05190821 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760447 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760446 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05151700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05274676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05861350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05838212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05620341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05620337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24760411 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10546957 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546954 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546959 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546958 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429865_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10546953 | - | - |
Mouse Genome 430 2.0 Array | 1429865_at | - | - |
Mouse Genome 430 2.0 Array | 1429865_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70979_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70979_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600022489.mm.1 | TC0600002928.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600022489.mm.1 | NM_027633.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600022489.mm.1 | NM_027633.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600022490.mm.1 | TC0600002928.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600022490.mm.1 | NM_027633.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600022490.mm.1 | NM_027633.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600022488.mm.1 | TC0600002928.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600022488.mm.1 | NM_027633.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600022488.mm.1 | NM_027633.3 | Refseq |
Mouse Expression Set 430 | 1429865_at | - | - |
Mouse Expression Set 430 | 1429865_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429865_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429865_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 416473 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103940035 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2710570 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2957167 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2957167 | - | - |
