Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400013907.mm.1 |
NM_199307.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013907.mm.1 |
NM_199307.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013907.mm.1 |
TC0400001714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013904.mm.1 |
NM_199307.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013904.mm.1 |
NM_199307.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013904.mm.1 |
TC0400001714.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013906.mm.1 |
NM_199307.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013906.mm.1 |
NM_199307.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013906.mm.1 |
TC0400001714.mm.1 |
MTA TC |