Assay Details
| Gene Symbol: Ece1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199307.2 | NP_955011.1 | 9 - 10 | 1196 | 93 | |
| GenBank mRNA | AK134088.1 | - | 9 - 10 | 1197 | 93 | |
| AK163790.1 | - | 6 - 7 | 1109 | 93 | ||
| AK205173.1 | - | 1 - 2 | 100 | 93 | ||
| BC060648.1 | - | 9 - 10 | 1136 | 6854311 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ece1
Genomic Map