Assay Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013807.2 | NP_038835.2 | 12 - 13 | 1598 | 80 | |
| GenBank mRNA | AK156754.1 | - | 12 - 13 | 1610 | 80 | |
| BC063051.1 | - | 12 - 13 | 1598 | 5718130 | 80 | |
| U21392.1 | - | 12 - 13 | 1394 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk3
Genomic Map