Assay Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013807.2 | NP_038835.2 | 4 - 5 | 658 | 93 | |
| GenBank mRNA | AK156754.1 | - | 4 - 5 | 670 | 93 | |
| AK209626.1 | - | 2 - 3 | 94 | |||
| BC031180.1 | - | 4 - 5 | 650 | 4988360 | 93 | |
| BC063051.1 | - | 4 - 5 | 658 | 5718130 | 93 | |
| U21392.1 | - | 4 - 5 | 505 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk3
Genomic Map