Assay Details
| Gene Symbol: Prdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133503.1 | - | 4 - 5 | 833 | 63 | |
| AK149344.1 | - | 4 - 5 | 724 | 63 | ||
| BC129801.1 | - | 4 - 5 | 843 | 63 | ||
| JX154081.1 | - | 4 - 5 | 666 | 63 | ||
| U08185.1 | - | 5 - 6 | 983 | 63 | ||
More Information
Set Membership: |
|

Prdm1
Genomic Map