Assay Details
| Gene Symbol: Prdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133503.1 | - | 5 - 6 | 1936 | 75 | |
| AK149344.1 | - | 5 - 6 | 1827 | 75 | ||
| BC129801.1 | - | 5 - 6 | 1946 | 75 | ||
| JX154081.1 | - | 5 - 6 | 1769 | 75 | ||
| U08185.1 | - | 6 - 7 | 2086 | 75 | ||
More Information
Set Membership: |
|

Prdm1
Genomic Map