Assay Details
| Gene Symbol: Prep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011156.2 | NP_035286.1 | 12 - 13 | 1659 | 63 | |
| GenBank mRNA | AB007631.1 | - | 12 - 13 | 1542 | 63 | |
| AK053816.1 | - | 12 - 13 | 1670 | 63 | ||
| AK076257.1 | - | 12 - 13 | 1658 | 63 | ||
| AK133875.1 | - | 12 - 13 | 1659 | 63 | ||
| AK141851.1 | - | 12 - 13 | 1668 | 63 | ||
| AK161528.1 | - | 12 - 13 | 1658 | 63 | ||
| AK163774.1 | - | 4 - 5 | 535 | 63 | ||
| AK163787.1 | - | 12 - 13 | 1671 | 63 | ||
| AK170570.1 | - | 12 - 13 | 1668 | 63 | ||
| BC012869.1 | - | 12 - 13 | 1659 | 3967219 | 63 | |
| BC050830.1 | - | 12 - 13 | 1652 | 6519113 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prep
Genomic Map