Assay Details
| Gene Symbol: Gmeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122992.1 | NP_001116464.1 | 4 - 5 | 369 | 75 | |
| NM_020273.2 | NP_064669.2 | 3 - 4 | 306 | 75 | ||
| GenBank mRNA | AF210433.1 | - | 2 - 3 | 267 | 75 | |
| AK134349.1 | - | 3 - 4 | 307 | 75 | ||
| BC046983.1 | - | 4 - 5 | 340 | 6477130 | 75 | |
| BU701415.1 | - | 2 - 3 | 153 | 75 | ||
| BY732360.1 | - | 4 - 5 | 344 | 75 | ||
| DV660282.1 | - | 2 - 3 | 224 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gmeb1
Genomic Map