Assay Details
| Gene Symbol: Gmeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122992.1 | NP_001116464.1 | 8 - 9 | 885 | 75 | |
| NM_020273.2 | NP_064669.2 | 7 - 8 | 822 | 75 | ||
| GenBank mRNA | AF210433.1 | - | 6 - 7 | 783 | 75 | |
| AK134349.1 | - | 7 - 8 | 823 | 75 | ||
| BC046983.1 | - | 8 - 9 | 856 | 6477130 | 75 | |
| BU701415.1 | - | 6 - 7 | 669 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gmeb1
Genomic Map