Assay Details
Gene Symbol: Chmp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026885.3 | NP_081161.1 | 4 - 5 | 669 | 74 | |
GenBank mRNA | AK005267.1 | - | 4 - 5 | 637 | 74 | |
BC012230.1 | - | 4 - 5 | 610 | 4024872 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
