Assay Details
| Gene Symbol: Eya3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010166.3 | NP_034296.2 | 10 - 11 | 980 | 149 | |
| NM_210071.2 | NP_997592.1 | 9 - 10 | 941 | 149 | ||
| NM_211357.2 | NP_997596.1 | 7 - 8 | 760 | 149 | ||
| GenBank mRNA | AK144653.1 | - | 9 - 10 | 932 | 149 | |
| BC052860.1 | - | 8 - 9 | 744 | 149 | ||
| BC063259.1 | - | 9 - 10 | 869 | 6406090 | 149 | |
| U61112.1 | - | 6 - 7 | 645 | 149 | ||
| U81604.1 | - | 10 - 11 | 948 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eya3
Genomic Map