Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400012522.mm.1 |
NM_010166.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012522.mm.1 |
NM_210071.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012522.mm.1 |
NM_211357.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012522.mm.1 |
TC0400001594.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400012521.mm.1 |
NM_010166.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012521.mm.1 |
NM_210071.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012521.mm.1 |
NM_211357.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012521.mm.1 |
TC0400001594.mm.1 |
MTA TC |