Assay Details
| Gene Symbol: Ppp1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290725.1 | NP_001277654.1 | 2 - 3 | 174 | 106 | |
| NM_146154.3 | NP_666266.1 | 2 - 3 | 174 | 106 | ||
| GenBank mRNA | AX775983.1 | - | 2 - 3 | 139 | 106 | |
| BC025479.1 | - | 2 - 3 | 136 | 5354224 | 106 | |
| BY257668.1 | - | 2 - 3 | 167 | 106 | ||
| BY293277.1 | - | 2 - 3 | 175 | 106 | ||
| CK625884.1 | - | 2 - 3 | 106 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r8
Genomic Map