Assay Details
| Gene Symbol: Sec14l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029937.2 | NP_001025108.1 | 2 - 3 | 231 | 77 | |
| GenBank mRNA | BC138982.1 | - | 2 - 3 | 231 | 8862004 | 77 |
| BC138985.1 | - | 2 - 3 | 231 | 8862007 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l3
Genomic Map