Assay Details
| Gene Symbol: Rnf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290472.1 | NP_001277401.1 | 5 - 6 | 1150 | 80 | |
| NM_001290473.1 | NP_001277402.1 | 4 - 5 | 1131 | 80 | ||
| NM_145355.5 | NP_663330.2 | 6 - 7 | 1269 | 80 | ||
| NR_110959.1 | 4 - 5 | 929 | 80 | |||
| GenBank mRNA | AK034108.1 | - | 6 - 7 | 639 | 80 | |
| AK052436.1 | - | 5 - 6 | 559 | 80 | ||
| AK053358.1 | - | 5 - 6 | 516 | 80 | ||
| AK075756.1 | - | 5 - 6 | 1151 | 80 | ||
| AK077638.1 | - | 5 - 6 | 481 | 80 | ||
| AK149640.1 | - | 5 - 6 | 502 | 80 | ||
| AK165895.1 | - | 575 | 80 | |||
| AK167455.1 | - | 5 - 6 | 467 | 80 | ||
| AK168904.1 | - | 5 - 6 | 504 | 80 | ||
| BC014812.1 | - | 5 - 6 | 481 | 3153870 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf185
Genomic Map