Assay Details
Gene Symbol: Nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008698.2 | NP_032724.1 | 3 - 4 | 278 | 57 | |
GenBank mRNA | AJ001260.1 | - | 3 - 4 | 278 | 57 | |
AK013140.1 | - | 3 - 4 | 286 | 57 | ||
AK031804.1 | - | 3 - 4 | 294 | 57 | ||
AK147786.1 | - | 4 - 5 | 555 | 57 | ||
BC010837.1 | - | 3 - 4 | 276 | 4242309 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
