Assay Details
| Gene Symbol: Rasl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145216.3 | NP_660251.1 | 2 - 3 | 523 | 73 | |
| GenBank mRNA | AF498251.1 | - | 2 - 3 | 345 | 73 | |
| AK008807.1 | - | 2 - 3 | 524 | 73 | ||
| AK044011.1 | - | 2 - 3 | 524 | 73 | ||
| BC107201.1 | - | 3 - 4 | 404 | 40051192 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561124 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561125 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561126 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10577 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105841 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27561105 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105841 | - | - |
| Whole Mouse Genome G4122A | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | AK008807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | AK044011.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | BC107200.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | BC107201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302942 | NM_145216.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113922 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105841 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09492315 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09448690 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09492314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09448685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09448681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561126 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561124 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561125 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27561137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27561205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09448688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09426877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09467551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09496831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09529875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09277644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09448690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09448693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09719904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09492316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09448683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09492311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08999746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09426870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27561120 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10374013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374015 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427975_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10374012 | - | - |
| Mouse Genome 430 2.0 Array | 1427975_at | - | - |
| Mouse Genome 430 2.0 Array | 1427975_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75668_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75668_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100000702.mm.1 | NM_145216.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000702.mm.1 | NM_145216.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000702.mm.1 | TC1100000061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000700.mm.1 | NM_145216.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000700.mm.1 | NM_145216.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000700.mm.1 | TC1100000061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000701.mm.1 | NM_145216.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000701.mm.1 | NM_145216.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000701.mm.1 | TC1100000061.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427975_at | - | - |
| Mouse Expression Set 430 | 1427975_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427975_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427975_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427975_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427975_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 832373 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6660450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2719702 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2719702 | - | - |
| mouse-6 v1.1 (Illumina) | scl41877.4.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41877.4.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2719702 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2719702 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41877.4.1_1-S | - | - |

Rasl10a
Genomic Map