Assay Details
| Gene Symbol: Thoc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172438.3 | NP_766026.1 | 5 - 6 | 506 | 97 | |
| GenBank mRNA | AK050734.1 | - | 5 - 6 | 506 | 97 | |
| AK173078.1 | - | 3 - 4 | 344 | 97 | ||
| BC020332.1 | - | 4 - 5 | 471 | 97 | ||
| BC021779.1 | - | 4 - 5 | 480 | 97 | ||
| BC039758.1 | - | 5 - 6 | 507 | 3995737 | 97 | |
More Information
Set Membership: |
|

Thoc5
Genomic Map