Assay Details
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077661.1 | NP_001071129.1 | 2 - 3 | 524 | 88 | |
GenBank mRNA | AK035580.1 | - | 2 - 3 | 520 | 88 | |
BC066994.1 | - | 2 - 3 | 405 | 5696000 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK035580.1, BC066994.1, NM_001077661.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
