Assay Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF076845.1 | - | 5 - 6 | 559 | 128 | |
| AK046152.1 | - | 5 - 6 | 566 | 128 | ||
| AK051839.1 | - | 4 - 5 | 991 | 128 | ||
| AK088403.1 | - | 5 - 6 | 569 | 128 | ||
| AK135922.1 | - | 5 - 6 | 549 | 128 | ||
| BC047460.1 | - | 5 - 6 | 576 | 128 | ||
| BC061249.1 | - | 5 - 6 | 595 | 30249148 | 128 | |
| Y16894.2 | - | 5 - 6 | 591 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hus1
Genomic Map