Assay Details
Gene Symbol: Sun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290519.1 | NP_001277448.1 | 5 - 6 | 414 | 84 | |
NM_001290520.1 | NP_001277449.1 | 3 - 4 | 337 | 84 | ||
NM_177576.3 | NP_808244.1 | 4 - 5 | 441 | 84 | ||
GenBank mRNA | AK052771.1 | - | 4 - 5 | 442 | 84 | |
AK132922.1 | - | 5 - 6 | 415 | 84 | ||
BC109334.2 | - | 3 - 4 | 251 | 40051782 | 84 | |
HQ141857.1 | - | 5 - 6 | 404 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
