Assay Details
| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145552.2 | NP_663527.2 | 2 - 3 | 265 | 92 | |
| GenBank mRNA | AK077532.1 | - | 2 - 3 | 235 | 92 | |
| AK146404.1 | - | 2 - 3 | 262 | 92 | ||
| BC003262.1 | - | 2 - 3 | 241 | 3501393 | 92 | |
| BY140435.1 | - | 2 - 3 | 266 | 92 | ||
| U69600.1 | - | 2 - 3 | 160 | 92 | ||
More Information
Set Membership: |
|

Gnl2
Genomic Map