Assay Details
| Gene Symbol: Azin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047522.1 | - | 4 - 5 | 631 | 95 | |
| AK077201.1 | - | 4 - 5 | 631 | 95 | ||
| AK148717.1 | - | 4 - 5 | 635 | 95 | ||
| BC138973.1 | - | 4 - 5 | 543 | 8861995 | 95 | |
| BC138974.1 | - | 4 - 5 | 543 | 8861996 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Azin2
Genomic Map