Assay Details
| Gene Symbol: Azin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047522.1 | - | 8 - 9 | 1272 | 96 | |
| AK077201.1 | - | 8 - 9 | 1272 | 96 | ||
| AK154564.1 | - | 8 - 9 | 1274 | 96 | ||
| BC065772.1 | - | 6 - 7 | 881 | 96 | ||
| BC138973.1 | - | 8 - 9 | 1184 | 8861995 | 96 | |
| BC138974.1 | - | 8 - 9 | 1184 | 8861996 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Azin2
Genomic Map