| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000002185.mm.1 |
NM_001122892.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002185.mm.1 |
NM_001122892.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002185.mm.1 |
NM_001122893.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002185.mm.1 |
NM_001122893.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002185.mm.1 |
NM_008054.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002185.mm.1 |
NM_008054.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002185.mm.1 |
TC1000000326.mm.1 |
MTA TC |