Assay Details
| Gene Symbol: Oprm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039652.2 | NP_001034741.1 | 5 - 6 | 73 | ||
| GenBank mRNA | AF062752.1 | - | 2 - 3 | 114 | 73 | |
| AF062753.1 | - | 5 - 6 | 1380 | 73 | ||
| AF074972.1 | - | 5 - 6 | 1215 | 73 | ||
| AF074973.1 | - | 4 - 5 | 1291 | 73 | ||
| AF074974.1 | - | 6 - 7 | 1500 | 73 | ||
| AF167568.1 | - | 7 - 8 | 1686 | 73 | ||
| AF260311.1 | - | 4 - 5 | 1039 | 73 | ||
| AK038389.1 | - | 5 - 6 | 73 | |||
| AY923182.1 | - | 4 - 5 | 1142 | 73 | ||
| BC119545.1 | - | 5 - 6 | 1380 | 40056952 | 73 | |
| DQ363374.1 | - | 5 - 6 | 882 | 73 | ||
| DQ868787.1 | - | 7 - 8 | 73 | |||
| DQ868788.2 | - | 7 - 8 | 73 | |||
| EF105311.1 | - | 7 - 8 | 73 | |||
| EF105312.1 | - | 7 - 8 | 73 | |||
| EF105313.1 | - | 8 - 9 | 73 | |||
| EF105314.1 | - | 6 - 7 | 73 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Oprm1
Genomic Map