Assay Details
| Gene Symbol: Gale | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178389.3 | NP_848476.1 | 6 - 7 | 881 | 77 | |
| GenBank mRNA | AK009083.1 | - | 7 - 8 | 749 | 77 | |
| AK146184.1 | - | 7 - 8 | 772 | 77 | ||
| BC027438.1 | - | 6 - 7 | 881 | 4224021 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gale
Genomic Map