Assay Details
| Gene Symbol: Sh3d21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162533.1 | NP_001156005.1 | 12 - 13 | 1026 | 61 | |
| NM_025856.3 | NP_080132.1 | 8 - 9 | 690 | 61 | ||
| GenBank mRNA | AK076962.1 | - | 8 - 9 | 655 | 61 | |
| AK155132.1 | - | 12 - 13 | 947 | 61 | ||
| BC019568.1 | - | 12 - 13 | 943 | 61 | ||
| BC039777.1 | - | 8 - 9 | 880 | 61 | ||
| BC053421.1 | - | 8 - 9 | 651 | 6706860 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076962.1 transcripts.
Set Membership: |
|

Sh3d21
Genomic Map