Assay Details
Gene Symbol: Pomgnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290658.1 | NP_001277587.1 | 9 - 10 | 1020 | 65 | |
NM_026651.2 | NP_080927.1 | 10 - 11 | 1106 | 65 | ||
NM_029786.2 | NP_084062.2 | 9 - 10 | 1007 | 65 | ||
GenBank mRNA | AB053220.1 | - | 10 - 11 | 1088 | 65 | |
AK002638.1 | - | 9 - 10 | 999 | 65 | ||
AK015478.1 | - | 9 - 10 | 1022 | 65 | ||
AK019640.1 | - | 9 - 10 | 1012 | 65 | ||
AK217580.1 | - | 1 - 2 | 38 | 65 | ||
BC011201.1 | - | 10 - 11 | 1029 | 4190368 | 65 | |
BC023265.1 | - | 7 - 8 | 693 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
