Assay Details
| Gene Symbol: Toe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026654.2 | NP_080930.1 | 4 - 5 | 372 | 76 | |
| GenBank mRNA | AK016889.1 | - | 362 | 76 | ||
| AK019829.1 | - | 4 - 5 | 372 | 76 | ||
| AK046576.1 | - | 4 - 5 | 403 | 76 | ||
| AK082975.1 | - | 4 - 5 | 353 | 76 | ||
| BC023109.1 | - | 4 - 5 | 348 | 4021435 | 76 | |
More Information
Set Membership: |
|

Toe1
Genomic Map