Assay Details
| Gene Symbol: Pdzrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018884.2 | NP_061372.2 | 5 - 6 | 1259 | 67 | |
| GenBank mRNA | AK143226.1 | - | 5 - 6 | 773 | 67 | |
| AK173098.1 | - | 5 - 6 | 1218 | 67 | ||
More Information
Set Membership: |
|

Pdzrn3
Genomic Map