Assay Details
| Gene Symbol: Rad18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC011120.1 | - | 7 - 8 | 929 | 4014013 | 116 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad18| Gene Symbol: Rad18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC011120.1 | - | 7 - 8 | 929 | 4014013 | 116 |
Set Membership: |
|