Assay Details
| Gene Symbol: Tcf7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079822.2 | NP_001073290.1 | 5 - 6 | 657 | 58 | |
| GenBank mRNA | BC128305.1 | - | 5 - 6 | 564 | 40109611 | 58 |
More Information
Updated Mapping Information
Set Membership: |
|

Tcf7l1
Genomic Map