Assay Details
| Gene Symbol: C1rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181344.5 | NP_851989.3 | 4 - 5 | 643 | 87 | |
| GenBank mRNA | AF456428.1 | - | 3 - 4 | 527 | 87 | |
| AK038659.1 | - | 4 - 5 | 609 | 87 | ||
| AK133495.1 | - | 4 - 5 | 622 | 87 | ||
| AK134207.1 | - | 4 - 5 | 644 | 87 | ||
| AK162497.1 | - | 4 - 5 | 598 | 87 | ||
| BC070472.1 | - | 4 - 5 | 606 | 30627947 | 87 | |
More Information
Set Membership: |
|

C1rl
Genomic Map