Assay Details
| Gene Symbol: Cyp4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010011.3 | NP_034141.3 | 2 - 3 | 365 | 90 | |
| GenBank mRNA | AB018421.1 | - | 2 - 3 | 359 | 90 | |
| AK002528.1 | - | 2 - 3 | 366 | 90 | ||
| BC010747.1 | - | 2 - 3 | 350 | 4242518 | 90 | |
| BC051049.1 | - | 2 - 3 | 351 | 4234581 | 90 | |
| X69296.1 | - | 339 | 90 | |||
| Gene Symbol: Cyp4a31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201640.3 | NP_964002.2 | 2 - 3 | 398 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid metabolic process
leukotriene B4 catabolic process
oxidation-reduction process
monooxygenase activity
iron ion binding
arachidonic acid monooxygenase activity
arachidonic acid epoxygenase activity
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
alkane 1-monooxygenase activity
heme binding
metal ion binding
leukotriene-B4 20-monooxygenase activity
arachidonic acid omega-hydroxylase activity
fatty acid metabolic process
leukotriene B4 catabolic process
oxidation-reduction process
monooxygenase activity
iron ion binding
arachidonic acid monooxygenase activity
arachidonic acid epoxygenase activity
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
alkane 1-monooxygenase activity
heme binding
metal ion binding
leukotriene-B4 20-monooxygenase activity
arachidonic acid omega-hydroxylase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024033 | NM_201640.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024033 | BC013476.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050628 | NM_201640.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952399 | NM_201640.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952399 | BC013476.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2803 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117587 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024033 | NM_201640.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024033 | BC013476.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050628 | NM_201640.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952399 | NM_201640.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952399 | BC013476.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224763 | - | - |
| Whole Mouse Genome G4122A | A_51_P254895 | BC013476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254895 | NM_010011.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254895 | BC013476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254895 | NM_010011.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P218449 | NM_201640.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P218449 | BC013476.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23225036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23225050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23225042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23225017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03953406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03615011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23225001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03576260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03576264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03618354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03618353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23225040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23225042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23225039 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507173 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507172 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440134_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424943_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507163 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507177 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507171 | - | - |
| Mouse Genome 430 2.0 Array | 1424943_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424943_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 666168_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 666168_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400008970.mm.1 | NM_010011.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008970.mm.1 | NM_010011.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008970.mm.1 | TC0400004187.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400009011.mm.1 | NM_201640.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009011.mm.1 | NM_201640.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009011.mm.1 | TC0400001229.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424943_at | - | - |
| Mouse Expression Set 430 | 1424943_PM_at | - | - |
| Mouse Expression Set 430 | 1424853_PM_s_at | - | - |
| Mouse Expression Set 430 | 1424853_s_at | - | - |
| Murine Genome U74v2 Set | 98353_at | - | - |
| Murine Genome U74v2 Set | 98353_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424853_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424943_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424943_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424853_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424943_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424943_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440134_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440134_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 704099 | - | - |
| ABI Mouse Genome Survey Microarray | 762919 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2529509 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2960044 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2960044 | - | - |

Cyp4a10
Genomic Map