Assay Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC047211.1 | - | 3 - 4 | 250 | 6392183 | 65 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC047211.1 transcripts.
Set Membership: |
|

Ccdc37
Genomic Map