Assay Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173775.3 | NP_776136.2 | 10 - 11 | 1118 | 65 | |
| GenBank mRNA | BC047211.1 | - | 8 - 9 | 800 | 6392183 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc37
Genomic Map