Assay Details
Gene Symbol: Ccdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173775.3 | NP_776136.2 | 11 - 12 | 1248 | 129 | |
GenBank mRNA | BC047211.1 | - | 9 - 10 | 930 | 6392183 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
