Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600020293.mm.1 |
TC0600002613.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020293.mm.1 |
NR_028081.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020293.mm.1 |
NR_028081.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020285.mm.1 |
TC0600002613.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020285.mm.1 |
NM_001163427.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020285.mm.1 |
NM_001163427.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020285.mm.1 |
NM_028505.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020285.mm.1 |
NM_028505.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020285.mm.1 |
NR_028081.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020285.mm.1 |
NR_028081.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020292.mm.1 |
TC0600002613.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020292.mm.1 |
NR_028081.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020292.mm.1 |
NR_028081.1 |
Refseq |