Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023077.mm.1 |
TC0600003001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023077.mm.1 |
NM_028335.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023077.mm.1 |
NM_028335.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023085.mm.1 |
TC0600003001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023085.mm.1 |
NM_028335.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023085.mm.1 |
NM_028335.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023075.mm.1 |
TC0600003001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023075.mm.1 |
NM_028335.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023075.mm.1 |
NM_028335.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023076.mm.1 |
TC0600003001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023076.mm.1 |
NM_028335.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023076.mm.1 |
NM_028335.2 |
Refseq |