Assay Details
| Gene Symbol: Zfp248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028335.2 | NP_082611.1 | 5 - 6 | 587 | 83 | |
| GenBank mRNA | AK012874.1 | - | 4 - 5 | 538 | 83 | |
| BC049087.1 | - | 2 - 3 | 105 | 83 | ||
| BC082571.1 | - | 5 - 6 | 587 | 30615600 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012874.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786879 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021231 | BC082571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021231 | NM_028335.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28544 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135960 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137922 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021231 | BC082571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021231 | NM_028335.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24786868 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137922 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135960 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06042698 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01429433 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06042676 | - | - |
| Whole Mouse Genome G4122A | A_51_P170050 | BC049087.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170050 | BC082571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170050 | AK012874.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170050 | NM_028335.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150951 | AK049763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150951 | AK084325.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135960 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137922 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05536039 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05536047 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05536040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24786877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24786790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05536040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05251297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05867995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05868000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05621171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05178842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24786885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05621188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05128110 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547281 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547277 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547275 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547280 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453531_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436473_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458267_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439534_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547274 | - | - |
| Mouse Genome 430 2.0 Array | 1458267_at | - | - |
| Mouse Genome 430 2.0 Array | 1458267_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439534_at | - | - |
| Mouse Genome 430 2.0 Array | 1436473_at | - | - |
| Mouse Genome 430 2.0 Array | 1453531_at | - | - |
| Mouse Genome 430 2.0 Array | 1453531_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436473_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439534_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72720_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72720_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600023074.mm.1 | TC0600003001.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600023074.mm.1 | NM_028335.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600023074.mm.1 | NM_028335.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600023073.mm.1 | TC0600003001.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600023073.mm.1 | NM_028335.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600023073.mm.1 | NM_028335.2 | Refseq |
| Mouse Expression Set 430 | 1458267_PM_at | - | - |
| Mouse Expression Set 430 | 1436473_at | - | - |
| Mouse Expression Set 430 | 1436473_PM_at | - | - |
| Mouse Expression Set 430 | 1439534_PM_at | - | - |
| Mouse Expression Set 430 | 1453531_at | - | - |
| Mouse Expression Set 430 | 1458267_at | - | - |
| Mouse Expression Set 430 | 1439534_at | - | - |
| Mouse Expression Set 430 | 1453531_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458267_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453531_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458267_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436473_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453531_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436473_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439534_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439534_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230997 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230997 | - | - |

Zfp248
Genomic Map