Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023010.mm.1 |
TC0600002994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023010.mm.1 |
NM_030165.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023010.mm.1 |
NM_030165.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023008.mm.1 |
TC0600002994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023008.mm.1 |
NM_030165.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023008.mm.1 |
NM_030165.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023006.mm.1 |
TC0600002994.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023006.mm.1 |
NM_030165.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023006.mm.1 |
NM_030165.3 |
Refseq |