Assay Details
Gene Symbol: Csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030165.3 | NP_084441.3 | 3 - 4 | 1259 | 74 | |
GenBank mRNA | AK019496.1 | - | 3 - 4 | 1262 | 74 | |
AK028045.1 | - | 3 - 4 | 1240 | 74 | ||
AK145992.1 | - | 3 - 4 | 1246 | 74 | ||
AK160867.1 | - | 3 - 4 | 1252 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
