Assay Details
| Gene Symbol: Rab43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039394.1 | NP_001034483.1 | 1 - 2 | 433 | 62 | |
| GenBank mRNA | AB232640.1 | - | 1 - 2 | 198 | 62 | |
| AJ534465.1 | - | 1 - 2 | 417 | 62 | ||
| AK010874.1 | - | 1 - 2 | 432 | 62 | ||
| AK158283.1 | - | 1 - 2 | 434 | 62 | ||
| BY753078.1 | - | 1 - 2 | 434 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab43
Genomic Map