Assay Details
| Gene Symbol: Grid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008167.2 | NP_032193.1 | 9 - 10 | 1348 | 126 | |
| GenBank mRNA | BC139823.1 | - | 9 - 10 | 1352 | 40141838 | 126 |
| D13266.1 | - | 9 - 10 | 1348 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grid2
Genomic Map