Assay Details
| Gene Symbol: Eogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175313.4 | NP_780522.1 | 8 - 9 | 1128 | 118 | |
| GenBank mRNA | AK037507.1 | - | 8 - 9 | 1129 | 118 | |
| AK199795.1 | - | 3 - 4 | 294 | 118 | ||
| BC048939.1 | - | 6 - 7 | 667 | 6439908 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eogt
Genomic Map